Find methods for your needs
Fast analysis of vitamin C using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of vitamin C. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 254 nm.| Market: | Food and Beverage |
| Keywords: | Ascorbic acid, HPLC, Hypersil Gold PFP, Uracil, UV detection, Vitamin C |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.