Find methods for your needs
Fast separation of neutrals from human plasma and urine using Thermo Scientific SOLA CX SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of neutrals from human plasma and urine. The samples are prepared using Thermo Scientific SOLA CX solid phase extraction (SPE) cartridges, and the separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with UV detection at 254 nm. For Research Use Only. Not for use in diagnostic procedures.| Market: | BioPharma |
| Keywords: | Accucore RP-MS, Corticosterone, HPLC, Human Plasma, Hydrocortisone, Neutrals, Progesterone, SOLA CX, SPE, Urine, UV detection |
| Matrix: | Human plasma, Urine |
For Research Use Only. Not for use in diagnostic procedures.