Find methods for your needs
Rapid and sensitive UHPLC screening for watersoluble vitamins in sports beverages
Description
• To demonstrate the capability of the Thermo Scientific™ Acclaim™ PolarAdvantage II (PA2) and Thermo Scientific™ Vanquish™ Flex Binary UHPLC system combination for the rapid separation of water soluble vitamins in sports beverages with excellent linearity, reproducibility, resolution, and recoveries. • To show the capability of the Vanquish Flex Binary UHPLC system to provide superior retention time precision through its novel pump features.| Market: | Food and Beverage |
| Keywords: | Ascorbic acid, Folic acid, Nicotinic acid, Rapid analysis, Water soluble vitamins, Acclaim PolarAdvantage II, Nicotinamide, UHPLC, beverage analysis, Cyanocobalamin, D-Pantothenic acid, Pyridoxine HCl, Vanquish Flex |
| Matrix: | Sports drinks |
| Author: | Jon Bardsley |
For Research Use Only. Not for use in diagnostic procedures.