 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
TN72225: Glycoprotein Monosaccharide Analysis using HPAE-PAD with Manually Prepared Eluent
Description
This work describes an HPAE-PAD method for monosaccharide composition analysis using manually prepared eluent. Monosaccharide analysis using electrolytically generated eluent has been described before. Here, three commercially available proteins, IgG, fetuin, and alpha-1-acid glycoprotein, were individually subjected to two hydrolysis conditions using 1)HCl, for the amino sugars galactosamine and glucosamine, and 2)TFA, for the neutral sugars mannose, glucose, and galactose. Results for method linearity, robustness, and accuracy are presented here. An ICS-6000 can be used for this application.| Market: | BioPharma | 
| Keywords: | Electrochemical detection, Glycoproteins, ICS-5000+, IgG, mAb, HPAE-PAD, CarboPac PA20 Column, Dionex AminoTrap column, Manual eluent preparation, Biopharmaceuticals, Quality control, ICS-6000, TN71, TN110, TN40, TN 72225, TN 20, TN 70671, TN20, TN70671 | 
| Matrix: | Glycoprotein acid digest | 
| Author: | Sachin Patil and Jeffrey Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA | 
For Research Use Only. Not for use in diagnostic procedures.