Find methods for your needs
Quantitation of Opiates to Low ng/mL Levels in Urine for Forensic Use Using an Affordable, High-Resolution, Accurate-Mass Mass Spectrometer
Description
Forensic toxicologists need an economical instrument capable of both screening a large number of compounds and quantifying smaller panels to industry-established limits. Here we present a method for quantitation of six opiates - morphine, codeine, hydromorphone, hydrocodone, oxymorphone, and oxycodone - in human urine down to low ng/mL levels. This work was performed on a Q Exactive Focus hybrid quadrupole Orbitrap mass spectrometer. For forensic use only.| Market: | Forensics; Toxicology |
| Keywords: | Codeine, drugs of abuse, opiates, Oxymorphone, Forensic Toxicology, Q Exactive Focus, morphine, hydromorphone, hydrocodone, oxycodone, PRM, parallel reaction monitoring |
| Matrix: | Urine |
| Author: | Kristine Van Natta, Marta Kozak |
| Affiliation: | Thermo Fisher Scientific |