Find methods for your needs
Essential Lipidomics Experiments Using the LTQ Orbitrap Hybrid Mass Spectrometer
Description
We tested the performance of a novel mass spectrometer, the Thermo Scientific LTQ Orbitrap, in a multitude of different applications in the field of lipidomics. The experiments presented here take advantage of the LTQ Orbitrap capabilities of multiple stage fragmentation, high resolution and mass accuracy to solve challenges of quantitating isomeric glycerophospholipids, resolving isobaric species, and elucidating fragmentation pathways and compositional assignment of various lipid species.| Market: | Food and Beverage |
| Keywords: | High resolution, LTQ Orbitrap, Accurate mass, Structural elucidation |
| Matrix: | Lipids |
| Author: | Thomas Moehring, Michaela Scigelova, Christer S. Ejsing, Dominik Schwudke, Andrej Shevchenko |
| Affiliation: | Max-Planck-Institute of Molecular Cell Biology and Genetics, Dresden, Germany; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.