Find methods for your needs
Separation of Acidic and Basic charge variants of Denosumab by Ion Exchange chromatography using pH Gradient Buffers
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Ion Exchange analysis of Denosumab. The separation was performed on a Thermo Scientific MAbPac SCX-10 and pH gradient buffers with a UV detection at 280 nm. With a run time of 50 minutes the method allows easy separation of charge variants of Denosumab.| Market: | BioPharma |
| Keywords: | HPLC, Denosumab, PDA detection, pH gradient, MabPac SCX-10, CX-1 pH Gradient Buffer A, CX-1 pH Gradient Buffer B |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.