Find methods for your needs
Dionex IonPac AS23-4µm Column Performance Test Using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. This record provides an eWorkflow for executing the QAR method on an Integrion HPIC system.| Market: | Environmental |
| Keywords: | Ion Chromatography (IC), Oxyhalides, Suppressed conductivity detection, EPA 300.1, Integrion HPIC, Quality Assurance Report (QAR), IonPac AS23-4um column, Dionex EGC 500 K2CO3, Dionex AERS 500 Carbonate suppressor |
| Matrix: | Water |
| Author: | Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.