Find methods for your needs
Accurately Monitor Protein PEGylation Reactions by HPLC-UV-CAD
Description
This application provides a fast and accurate way for biopharmaceutical chemists to monitor PEGylation reactions by using HPLC with complementary UV/Vis and charged aerosol detection.Separation on the MAbPac RP resolves PEGylation reagents, unreacted IgG, and PEGylated IgG products that elute as a broad hump.| Market: | BioPharma |
| Keywords: | protein characterization, Universal Detection, Protein Conjugation |
| Matrix: | PEGylation reaction solution |
| Author: | Dave Thomas |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.