Find methods for your needs
AU203: Determination of Trace Concentrations of Oxyhalides and Bromide in Municipal and Bottled Waters Using a Compact Ion Chromatography System
Description
To demonstrate that oxyhalides and bromide can be run successfully using a Thermo Scientific™ Dionex™ Integrion™ HPIC™ system combined with a Thermo Scientific™ Dionex™ IonPac™ AS19-4μm column, featuring smaller resin particles, to achieve faster analysis without compromising data quality compared to using the Dionex IonPac AS19 column as in Application Note 167.| Market: | Environmental |
| Keywords: | Bromate, Disinfection Byproducts, suppressed conductivity detection, Drinking water, Dionex IonPac AS19-4μm column, AN167, AU203, US EPA 300.1 Part B, TN175, AU 203 |
| Matrix: | Water |
| Author: | Jingli Hu; Jeffrey Rohrer |
| Affiliation: | ThermoFisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.