Find methods for your needs
Analysis of Diquat and Paraquat Using UHPLC Orbitrap MS - Method Development, Matrix Effects and Performance
Description
We investigate the effect of UHPLC mobile phases and operational parameters of a Orbitrap MS system used in the analysis of quaternary ammonium herbicides paraquat (PQ) and diquat (DQ). This includes separation optimization on a Thermo Scientific Acclaim Trinity Q1 column which was specifically designed for this application. Based on results obtained from this study, a method was developed for the unambiguous identification of PQ and DQ in environmental water samples with the ability to deliver analytical data with superior SNR, high precision and accuracy.| Market: | Environmental |
| Keywords: | Herbicides, UHPLC, Water analysis, Orbitrap MS, Acclaim Trinity Q1 Analytical Column, U.S. EPA Methods, Ontario Regulation 169/03 |
| Matrix: | Water |
| Author: | Jonathan Beck, Paul Yang, Xiaodong Liu, Chunyan Hao, Xiaoming Zhao, Franca Morra, Charles Yang, Maciej Bromirski, and Dipankar Ghosh |
| Affiliation: | Ontario Ministry of the Environment - Canada; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.