Find methods for your needs
AU200: Fast Anion Determinations in Environmental Waters Using a High-Pressure Compact Ion Chromatography System
Description
The determination of anions is one of the most important IC applications. Here we have updated Application Note 154 with the 4µm-resin-particle IonPac AS18-Fast-4µm column with expected backpressures approaching the previous system's limitations. In this Application Update, fast anion runs (9 min) were achieved with a gradient separation, saving 4 min run time with while retaining similar chromatography with higher sensitivity than the previous application. This analysis was facilitated by the high pressure capable Dionex Integrion HPIC system configured for Reagent-Free IC.| Market: | Environmental |
| Keywords: | Environmental water analysis, Fast IC, surface water, IonPac AS18, Suppressed Conductivity, EPA 300.1, wastewater, Drinking water, Integrion, municipal drinking water, IonPac AS18-Fast-4µm column, Integrion HPIC, Integrion RFIC |
| Matrix: | Municipal wastewater |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.