Find methods for your needs
Determination of Potential Chloride and Sulfate in Denatured Ethanol Samples Using a Compact Ion Chromatography System
Description
This method shows the direct determination of potential chloride and sulfate in a peroxide-treated denatured ethanol sample, previously published in Application Update 194. In this record, the method is executed on an Integrion IC system.| Market: | Chemical |
| Keywords: | biofuel, direct injection, Ethanol, HPIC, anions, IC, Chloride, Sulfate, Suppressed Conductivity, IonPac AS22 Column, carbonate eluent, Integrion, Ion chromatograhy, Denatured alcohol, AERS 500 carbonate suppressor, potential chloride, Potential sulfate, existent chloride, existent sulfate, ASTM D7319-13, Integrion HPIC, Denatured ethanol |
| Matrix: | Peroxide treated denatured ethanol |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.