Find methods for your needs
Determination of Galactosamine Containing Organic Impurities in Heparin Using a Compact Ion Chromatography System
Description
This record shows that AN233 can be run using an Integrion ion chromatography system. In AN233, the organic impurities in heparin are determined by HPAE-PAD using the Dionex CarboPac PA20 column following the USP monograph method. This method provides sensitive determination of galactosamine in acid-hydrolyzed heparin samples, enabling the identification of heparin that has been contaminated with chondroitin sulfates.| Market: | BioPharma |
| Keywords: | heparin, USP, ion chromatography, HPAE-PAD, CarboPac PA20 Column, Integrion, TN 20, TN 70671, TN70671, N20, AN233, TN176 |
| Matrix: | Water |
| Author: | Jingli Hu; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.