Find methods for your needs
TN62: Reducing Carbonate Interference in Anion Determinations with the Carbonate Removal Device (CRD): Carbonated Mineral Water
Description
Carbonate can interfere with accurate quantification of adjoining anions, such as sulfate. Here we show the efficient removal of the carbonate peak from a carbonated mineral water sample using the Dionex CRD 200 device.| Market: | Food and Beverage |
| Keywords: | Chlorate, Formate, Carbonate, IonPac AS18 column, fluoride, anions, RFIC, IC, Chloride, Acetate, Sulfate, Suppressed Conductivity, Bromide, CRD 200 device, carbonated mineral water, carbonate removal |
| Matrix: | Carbonated mineral water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.