Find methods for your needs
AN1142: Determination of Aluminum in OTC Pharmaceutical Products
Description
Aluminum-containing compounds are used extensively in cosmetics, prescription pharmaceuticals, and OTC drug products. In this application note, we report the validation of an IC method for the determination of aluminum in OTC formulations using a Dionex IonPac CS10 analytical column and postcolumn derivatization with Tiron, followed by UV absorbance detection at 310 nm. The method was evaluated in terms of linearity, precision, accuracy, ruggedness, and limit of quantitation for aluminum. The method was evaluated with antacids and deodorants. An ICS-6000 can be used for this application.| Market: | Pharma |
| Keywords: | Antacids, UV Absorbance, Aluminum, Postcolumn derivatization, Dionex IonPac CS10 Column, Dionex ICS-5000+ Reagent-Free HPIC System, Antiperspirants, Tiron, OTC formulations, Pharmaceutical products, aluminum chloride, Aluminum chlorohydrate, ICS-6000 |
| Matrix: | Antacids, Deodorants |
| Author: | Manali Aggrawal, Jeff Rohrer |
| Affiliation: | Thermo Fisher Sientific |
For Research Use Only. Not for use in diagnostic procedures.