Find methods for your needs
AU196: Anion Determinations in Municipal Drinking Water Samples Using EPA Method 300.1 (A) on an Integrated IC System
Description
Four California municipal drinking water samples were analyzed according the U.S. EPA Method 300.1 (A) on a 4 x 250 mm IonPac AS22 column using manually prepared carbonate-based eluent. Six inorganic anions were detected.| Market: | Environmental |
| Keywords: | Bromide, acetate, anions, carbonate eluent, chloride, formate, IonPac AS22 column, nitrate, nitrite, phosphate, sulfate, manually prepared eluent |
| Matrix: | Municipal drinking water |
| Author: | T.Christison, C.Tanner, J.E.Madden, L. Lopez |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.