Find methods for your needs
AU175: Determination of Organic Acids and Inorganic Anions in Lithium-Containing Boric Acid-Treated Nuclear Power Plant Waters
Description
In nuclear power plants with pressurized water reactors the primary coolant includes boric acid, which at high temperature and pressure can form crud. Lithium hydroxide is added to prevent this. Trace anionic impurities can serve as catalysts for corrosion. The experimental approach shown in this work combines the direct injection, electrolytically generated tetraborate eluent approach to analyze lithium-containing borated waters for fluoride, formate, chloride, and sulfate with the addition of a continuously regenerated cation trap column to remove lithium and other cations from the sample.Market: | Other |
Keywords: | Formate, IonPac AS22, fluoride, boron, Chloride, Acetate, Sulfate, boric acid, crud, lithium hydroxide, Nuclear Power Plant, pressurized water reactor, tetraborate |
Matrix: | Boric acid-treated water |
Author: | Rich Kornfeld, Brian De Borba, and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.