Find methods for your needs
AU145: Determination of Perchlorate in Drinking Water by Ion Chromatography
Description
In this updated method, the perchlorate anion is determined in 15 min by using a 2 mm Dionex IonPac AS16 column, EG50-generated hydroxide eluent, a 1000 μL injection, and suppressed conductivity detector. The resulting single-operator method detection limit (MDL) for perchlorate in a simulated high-ionic-strength water sample was 0.10 μg/L after sample preparation.| Market: | Water Analysis |
| Keywords: | Drinking Water, Ground water, RFIC, AU 145, EPA Method 314.0, IonPac AS16, OnGuard |
| Matrix: | Water |
| Author: | David H. Thomas and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.