Find methods for your needs
AN1078: Determination of Benzenesulfonic Acid Counterion in Amlodipine Besylate by Ion Chromatography
Description
This study demonstrates the determination of benzenesulfonate in amlodipine besylate using an electrolytically generated potassium hydroxide eluent with a hydroxide-selective Dionex IonPac AS18 anion exchange column and suppressed conductivity detection. This method provides a simple and reliable approach to determine the counter ion in the drug substance.Market: | Pharma |
Keywords: | Calcium Channel Blocker, USP monograph, RFIC, AN 1078, Dionex IonPac AS18, Amlodipine besylate |
Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.