Find methods for your needs
AN178: Improved Determination of Trace Concentrations of Perchlorate in Drinking Water Using Preconcentration with Two-Dimensional IC and Suppressed Conductivity Detection
Description
This application note describes a two-dimensional system for determining trace concentrations of perchlorate in environmental waters. The method resulted in an improvement to the existing EPA Methods 314.0 and 314.1 by providing lower detection limits and improved precision and recovery of perchlorate fortified in different sample matrices. In addition, samples can be injected directly without the need for sample preparation, a sample rinse step with sodium hydroxide, and the addition of matrix ions to the standards and samples.| Market: | Environmental |
| Keywords: | AN 178, EPA Method 314.0, EPA Method 314.1, IonPac AS16, IonPac AS20, IonPac TAC-ULP1, EPA 314.2 |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.