Description
The Thermo Scientific HPLC system is applied for the analysis of glucosamine sulfate. The separation is performed on a Thermo Scientific Hypercarb column in less than 5 minutes, with MS detection.| Market: | BioPharma |
| Keywords: | Glucosamine sulfate, HPLC, Hypercarb, LC-MS, MS detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.