Description
Size exclusion chromatography (SEC) is one of the major analyses in biotherapeutics characterization. Here we show a MAbPac SEC-1 column test with the Thermo Scientific Vanquish Flex UHPLC system.| Market: | Other |
| Keywords: | Monoclonal antibody, protein characterization, Size exclusion, Size exclusion chromatography, Peptide |
| Matrix: | 50 mM Sodium phosphate pH 6.8 + 300 mM NaCl |
For Research Use Only. Not for use in diagnostic procedures.