Description
Hydrophobic interaction liquid chromatography (HIC) is one of the major analyses in biotherapeutics characterization. Here we show a high resolution separation of proteins and a mAb in 20 minutes with the Thermo Scientific Vanquish Flex UHPLC system. The unique chemistry of the MAbPac HIC-20 column allows the analysis of hydrophilic proteins such as myoblogin and ribonuclease A as well as more hydrophobic mAbs.| Market: | BioPharma |
| Keywords: | protein characterization, MAbPac, Monoclonal antibody, Hydrophobic interaction liquid chromatography |
| Matrix: | 100 mM sodium phosphate, 2 M ammonium sulfate, pH 7 |
| Author: | Michael Heidorn |
For Research Use Only. Not for use in diagnostic procedures.