Find methods for your needs
AN1121: Ion Chromatography Assay for Lithium in Lithium Citrate
Description
This application note describes an ion chromatography (IC) based method that uses a Thermo Scientific™ Dionex™ IonPac™ CS12A-5µm cation-exchange column, an electrolytically generated MSA eluent, and suppressed conductivity detection to determine lithium in lithium citrate. The method proposed in this application note was validated following the guidelines outlined in USP General Chapter <1225>, Validation of Compendial Procedures.| Market: | Pharma |
| Keywords: | USP monograph, Dionex IonPac CS12A-5μm column, RFIC, AN 1121, Lithium citrate, Biopolar disorder |
| Matrix: | Water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.