Find methods for your needs
AN2967: Implementation of a Walk-Up High-Pressure Capillary Ion Chromatograph for the Fast Separation of Pharmaceutical Relevant Inorganic Anions (allergy tablet)
Description
The API counter ion impacts the drug’s efficacy, solubility, and toxicity, and therefore counter ion determinations are important to pharmaceutical companies during all stages of product development and as part of product quality monitoring. In this example, the chloride counter ion is determined in a water extract of a pharmaceutical drug tablet designed for the treatment of seasonal allergic reactions. Here fast chloride determinations were demonstrated on IonSwift MAX-200 capillary column using a walk-up high pressure capillary IC system.| Market: | Pharma |
| Keywords: | allergy tablet, cap IC, capillary IC, counter anions, counter ions, fast IC, IonSwift MAX-200 column, monolith |
| Matrix: | Water solution of a pharmaceutical drug tablet developed for treatment of seasonal allergy symptoms |
| Author: | T. Christison, J.E. Madden, F. Pang, and K. Divan |
For Research Use Only. Not for use in diagnostic procedures.