Description
One of the most important applications of IC in clinical chemistry is the analysis of physiological fluids such as urine, plasma, and serum both for inorganic and organic anions and cations. This application note outlines the analysis of physiological fluids using ion chromatography. Here shows the cation method, the IonPac CS12A is coupled with the Cation Self-Regenerating Suppressor (CSRS) provides fast, isocratic separations of lithium, sodium, ammonium, potassium, magnesium, and calcium in less than 12 minutes using a dilute sulfuric acid eluent.Market: | Pharma |
Keywords: | Ammonium, Calcium, Cations, IC, IonPac CS12A column, lithium, Magnesium, Physiological Fluids, plasma, Potassium, Sodium, urine |
Matrix: | Physiological fluids |
For Research Use Only. Not for use in diagnostic procedures.