Find methods for your needs
AN151: Determination of Perchlorate in Environmental Waters by Ion Chromatography Coupled with Electrospray Mass Spectrometry (IC-MS) Be the first to rate this application
an151 determination perchlorate environmental waters by ion chromatography coupled with electrospray mass spectrometry icms
Description
This application note describes the use of IC-MS to determine perchlorate in environmental waters. The described method includes in-line diversion of the sample matrix to waste and the use of an internal standard. Quantification using 99 m/z as compared to the perchlorate isotope (101 m/z) is also shown.
Market: Environmental
Keywords: EPA, Ground water, AN 151, AXP-MS, RFIC, IonPac AS16, Drinking water, EPA 332, US EPA Method 332
Matrix: Water
Author: Rosanne Slingsby and Chris Pohl
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA, USA
Uploaded on 4/30/2015.
For Research Use Only. Not for use in diagnostic procedures.