Find methods for your needs
Fast analysis of glutamate in beer by automated precolumn OPA derivatization
Description
The Thermo Scientific HPLC system is applied for the analysis of glutamate in beer. The sample preparation is automated, and the separation is performed using a Thermo Scientific Acclaim PolarAdvantage column, with UV detection at 330 nm. Chromatogram A is beer and chromatogram B is beer spiked with 250 µg/mL glutamic acid.| Market: | Food and Beverage |
| Keywords: | Acclaim PolarAdvantage, Automated sample preparation, Beer, Glutamate, HPLC, OPA derivatization, UV detection |
| Matrix: | Beer |
For Research Use Only. Not for use in diagnostic procedures.