Find methods for your needs
Determination of Steviosides by HPLC-CAD on UltiMate 3000
Description
This application combines the Acclaim Trinity P1 with charged aerosol and diode array detection to quantify stevioside sweeteners used in foods and beverages. The two detectors are complimentary at higher concentrations of steviosides, but under the conditions used here the charged aerosol detector provides superior sensitivity for most of the analytes.| Market: | Food and Beverage |
| Keywords: | Artificial sweeteners, Charged aerosol detector, Food and Beverage, Universal Detection, mixed mode separation |
| Matrix: | Standards in water |
| Author: | Dave Thomas |
| Affiliation: | Thermo Scientific |
For Research Use Only. Not for use in diagnostic procedures.