Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of sugars in cola drink. The separation is performed on a Thermo Scientific Accucore 150-Amide-HILIC HPLC column, with RI detection.| Market: | Food and Beverage |
| Keywords: | Accucore 150-Amide-HILIC, Cola, Fructose, Glucose, HPLC, RI detection, Sugars |
| Matrix: | Cola soft drink |
For Research Use Only. Not for use in diagnostic procedures.