Find methods for your needs
AN104: Analysis of Personal Care Products by Ion Chromatography
Description
Ion chromatography (IC) with conductivity detection complements traditional HPLC-UV pharmaceutical separations. IC is especially useful when the analytes do not contain chromophores, or when it is desirable to determine chromophoric and nonchromophoric molecules in a single run. This application note addresses the use of IC for the analysis of personal care products, including compounds found in some toothpastes, hair permanent solutions, and acne medications. Several examples illustrate the variety of separations that can be achieved using IC.| Market: | Other |
| Keywords: | Chloride, Excipients, Organic acid, Orthophosphate, Salicylate, Salicylic acid, Citrate, sulfate, fluoride, acne medication, IonPac AS11, Person care products, Toothpaste, waving solution, Monofluorophosphate, Pyrophosphate, Trimetaphosphate, Tripolyphosphate |
| Matrix: | Toothpaste |
For Research Use Only. Not for use in diagnostic procedures.