Find methods for your needs
AN166: Application of Eluent Generation for Trace AnionAnalysis of Borated Waters
Description
Boron as boric acid is used to control the nuclear reaction in pressurized water reactors because of its strong neutron-absorbing characteristics. To prevent corrosion, anionic contamination is monitored. Here sub-ppb concentrations of inorganic anions are determined in borated waters (<7500 mg/L boron) by large volume, direct injection using a tetraborate gradient on IonPac AS14 column. The tetraborate eluent is automatically generated inline by titrating 50 mM boric acid with electrolytically generated KOH. This is an improvement over using manually prepared solutions (AU102, AU191).| Market: | Water Analysis |
| Keywords: | Chloride, Pressurized water reactors, sulfate, fluoride, borated power plant water, boron, power plant, trace anions, Nitrate, PWR, borate |
| Matrix: | Borated power plant water |
| Author: | Ed Kaiser and Jeff Rohrer |
For Research Use Only. Not for use in diagnostic procedures.