Description
The Thermo Scientific Acclaim Surfactant Plus column and charged aerosol detection are powerfully combined in the Thermo Scientific Vanquish UHPLC system to quickly characterize a complex mixture of chelants, ionic and non-ionic surfactants from a multipurpose alkaline cleaning concentrate. Because the charged aerosol detector can quantify any non-volatine analyte separated by HPLC, this method could be made quantitative for use in cleaning validation studies.| Market: | Pharma |
| Keywords: | Charged aerosol detector, Universal Detection, Vanquish UHPLC, Surfactants |
| Matrix: | 50:50 acetonitrile:deionized water |
| Author: | Dave Thomas |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.