Find methods for your needs
AN1006: Determination of Carbohydrates in Urine by HPAE-PAD: Method 2
Description
The second method in this application note takes advantage of the selectivity and high capacity of the Dionex CarboPac MA1 column. Use of this column allows mannitol to be strongly retained with minimal interferences. These advantages have led to many publications that can be found in the reference section of the application note.Market: | Clinical Research |
Keywords: | carbohydrate, Lactulose, rhamnose, xylose, CarboPac MA1 column, Lactose, 3-O-methylglucose, mannitol, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 1006 |
Matrix: | Urine |
Author: | Deanna Hurum and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.