 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
AN1006: Determination of Carbohydrates in Urine by HPAE-PAD: Method 2
                                Description
The second method in this application note takes advantage of the selectivity and high capacity of the Dionex CarboPac MA1 column. Use of this column allows mannitol to be strongly retained with minimal interferences. These advantages have led to many publications that can be found in the reference section of the application note.| Market: | Clinical Research | 
| Keywords: | carbohydrate, Lactulose, rhamnose, xylose, CarboPac MA1 column, Lactose, 3-O-methylglucose, mannitol, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 1006 | 
| Matrix: | Urine | 
| Author: | Deanna Hurum and Jeffrey Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA | 
For Research Use Only. Not for use in diagnostic procedures.