Find methods for your needs
TN130: Fast Determinations of Inorganic Anions in Salton Sea Samples Using a High-Pressure IC System
Description
Monitoring salinity levels in estuarine ecosystems is critical to preventing salinity-specific diseases, especially in a saline lake, such as the Salton Sea with higher salt content than the ocean. This inland sea is being monitored for reclamation efforts. This application demonstrates fast anion and cation analysis of high saline samples, such as the Salton Sea, using a high pressure IC system and 2 mm fast separation columns. An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | Fast IC, IonPac AS18-4μm, HPIC, ICS-5000+ HPIC system, Salton Sea, fluoride, anions, Nitrate, RFIC, saline, Chloride, Sulfate, ICS-6000 |
| Matrix: | Saline lake |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.