Find methods for your needs
TN135: Determinations of Monosaccharides and Disaccharides in Beverages by Capillary HPAE-PAD
Description
Mono- and disaccharide sugar determinations are often used in the food and beverage industry to ensure the quality of a formulated product, to maintain or select for desired sweetness, and to characterize and confirm the source of the carbohydrates (cane, beet, or corn). This application demonstrates the determinations of high concentrations of sugars in beverages using a capillary IC system. Sugars were detected by HPAE-PAD with 4-potential waveform and Au on PTFE working electrode and 0.015” thick gasket.| Market: | Food and Beverage |
| Keywords: | Glucose, Carbonated beverage, Mannose, Sucrose, fructose, galactose, HPAE-PAD, capillary, CarboPac PA20 Column, TN 20, TN 70671, TN20, TN70671, TN136, Capillary HPAE-PAD:TN 135, AN82 |
| Matrix: | Carbonated beverage |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.