Find methods for your needs
AU137: Determination of Trace Lithium in Industrial Process Waters
Description
In a variety of situations, carryover from one body of water to another is measured by monitoring a tracer compound. Lithium is a good choice for a tracer because it is very soluble and has a low background concentration in public water supplies. This Application Update describes a rapid and sensitive ion chromatographic method for the determination of trace lithium using a microbore IonPac CS12A column.| Market: | Water Analysis |
| Keywords: | Ethanolamine, Preconcentration, Tracer, Hydrazine, TCC-LP1, CG10, Process water, Activated sludge reactor, morpholine, Lithium, Sodium, TN 73892, TN73892, AU 137 |
| Matrix: | Water |
| Author: | Edward Kaiser and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.