Find methods for your needs
Purity and mass determination using Thermo Scientific Accucore aQ HPLC columns: a column lifetime study
Description
A fast and reliable separation for quality control of small molecule drug candidates is demonstrated using Thermo Scientific Accucore aQ HPLC columns. The method run time was 3.25 minutes with each compound adequately resolved. The excellent peak shape was maintained for more than 11000 injections.| Market: | Other |
| Keywords: | Accucore aQ, Column lifetime, HPLC, Hydrocortisone, Ketoprofen, Sulfamerazine, Sulfamethoxazole, Sulfanilamide, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.