Find methods for your needs
Monitoring protein deamidation using a Thermo Scientific ProPac WCX-10 column
Description
The Thermo Scientific UltiMate 3000 Titanium HPLC system is applied to monitor the products of protein deamidation. The separation is performed on a Thermo Scientific ProPac WCX-10 column, with UV detection at 280 nm.| Market: | BioPharma |
| Keywords: | HPLC, ProPac WCX-10, Protein deamidation, RNase A, UV detection |
| Matrix: | Ribonuclease A (RNase A) |
For Research Use Only. Not for use in diagnostic procedures.