Description
In this Application Update, ion chromatography coupled with pulsed amperometric detection is used to determine iodide in urine. This method is specific, sensitive, and rapid. Sample preparation involves removal of urine components with molecular weights greater than 10,000 daltons, sample dilution, and treatment with a polymeric, reversed-phase cartridge (OnGuard™-RP).| Market: | Clinical Research |
| Keywords: | electrochemical detection, halide, hyperthyrosis, IC, IC-PAD, iodide, ion chromatography, IonPac AS11, OnGuard RP, PAD, thyroid disfunction |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.