Find methods for your needs
AN1105: Determination of Anions and Cations in Produced Water from Hydraulic Fracturing
Description
This Application Note describes the quantification of anions and cations in produced water using a Thermo Scientific Dionex ICS-5000+ Reagent-Free High-Pressure Ion Chromatography (HPIC) system with a Thermo Scientific™ Dione IonPac CS16 capillary column and Thermo Scientific Dionex ICS-2100 Integrated Reagent-Free IC (RFIC) system with a Dionex IonPac AS18 microbore column. An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | Dionex ICS-2100, Dionex ICS-5000+, Dionex IonPac AS18, Dionex IonPac CS16, Formate, Fracking, fluoride, RFIC, Chloride, Acetate, Sulfate, capillary IC, Bromide, ICS-6000 |
| Matrix: | Wastewater |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.