Find methods for your needs
AU150: Determination of Plant-Derived Neutral Oligo- and Polysaccharides
Description
Since the publication of Application Note 67, Dionex introduced the Dionex CarboPac™ PA200, a column designed for even higher-resolution separations than the Dionex CarboPac PA1 or PA100 columns. The improved resolution is due in part to the PA200’s significantly smaller substrate (5.5 μm) and latex (43 nm) beads. These and other changes produce a column that has greater resolution and requires a lower sodium acetate concentration to elute a given oligosaccharide than the PA100. The AU highlights the benefits of the PA200 for these applications.| Market: | Food and Beverage |
| Keywords: | amylopectins, amylose, Carbohydates, CarboPac PA200 column, Electrochemical detection, fructans, inulins, Maltodextrins, Polysaccharides, oligosaccharides, HPAE-PAD, TN71, TN 20, TN 70671, TN20, TN70671, AU 150 |
| Matrix: | Plants |
| Author: | David H. Thomas and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.