Find methods for your needs
An accurate related substances analysis of atorvastatin and ezetimibe using HPLC-UV
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the related substance analysis of atorvastatin and ezetimibe. The separation is performed on a Thermo Scientific Accucore PFP HPLC column, with UV detection at 260 nm.| Market: | Pharma |
| Keywords: | Accucore PFP, Atorvastatin, Ezetimibe, HPLC, Impurities, Related substances, UV detection |
| Matrix: | Formulation |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.