Find methods for your needs
AN1094: Determination of Cations in Hydraulic Fracturing Flowback Water from the Marcellus Shale
Description
This Application Note describes the quantification of cations in hydraulic fracturing flowback water from the Marcellus Shale using the Thermo Scientific Dionex ICS-5000+ Reagent-Free High-Pressure Ion Chromatography (HPIC) system with the Thermo Scientific Dionex IonPac CS16 standard bore (5 mm i.d.) and capillary (0.5 mm i.d.) columns. An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | Barium, Fracking, ICS-5000+, IonPac CS16, Magnesium, Strontium, Lithium, RFIC, Potassium, ion chromatography, Calcium, Sodium, capillary IC, Ammonium, ICS-6000 |
| Matrix: | Wastewater |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.