Find methods for your needs
SOLAμ for pre-analysis sample concentration to improve sensitivity
Description
This work demonstrates the use of Thermo Scientific SOLAμ SPE product to enhance sample pre-concentration prior to analysis. Additional benefits include reduced workflow and improved stability for analytes susceptible to loss or degradation during evaporation and reconstitution. The use of a Thermo Scientific Accucore HPLC column provided fast and efficient separation without the need for an ultra high pressure system. MS/MS detection was performed on a Thermo Scientific TSQ Vantage mass spectrometer. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | LC-MS/MS, matrix effects, micro elution, niflumic acid, no dry down, reproducibility, sample concentration, sensitivity, SOLAμ, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.