Find methods for your needs
Fast separation of impurities in aceclofenac solid dosage formulation using a reversed-phase HPLC-UV method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the fast separation of the impurities present in the nonsteroidal anti-inflammatory drug aceclofenac in solid dosage formulation. The separation was performed in less than 16 minutes using a Thermo Scientific Syncronis C18 5 µm column with UV detection at 275 nm.| Market: | Pharma |
| Keywords: | Aceclofenac, Diclofenac, Formulation, HPLC, NSAID, Syncronis C18, UV detection, Impurities |
| Matrix: | Formulation |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.