Find methods for your needs
Four high resolution peptide maps using a Thermo Scientific Acclaim PolarAdvantage II HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of ovalbumin, fetuin, β-casein and bovine serum albumin. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II HPLC column, with UV detection at 210 nm.| Market: | BioPharma |
| Keywords: | Acclaim PolarAdvantage II (PA2), Bovine serum albumin, fetuin, HPLC, Ovalbumin, Peptides, UV detection, β-casein |
| Matrix: | Protein |
For Research Use Only. Not for use in diagnostic procedures.