Description
This shows the determination of xylose in serum by HPAE-PAD. During the xylose absorption test, which is administered to evaluate small intestine absorption, the amount of xylose in serum is determined. If there is a bacterial infection of the small intestine, xylose is incompletely absorbed and is present in serum.| Market: | Clinical Research |
| Keywords: | Glucose, xylose, HPAE-PAD, TN71, TN 71, TN 20, TN 70671, TN20, TN70671, AU 125 |
| Matrix: | Serum |
| Author: | Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.